CE International Trading Mesh Screen, SS, 0.063 mm Sieve Opening, 250 mesh, 18" dia
Representative image only.
CE International Trading Mfr # LS450S005-#250MESH – Item # EW-59979-65
Mesh screens separate oversized particles from your sample
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)230
  • Tyler Standard Screen Size (mesh)250
  • Sieve Opening (mm)0.063
  • Outside Diameter (in)18
  • DescriptionMesh Screen, SS, 0.063 mm sieve opening, 250 mesh
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).