Celite 545 Filter Aid, Powder, 25 lb

Representative image only.
Celite Mfr # C21225LB – Item # EW-88068-91
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on

PRODUCT OPTIONS

$1,086.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaCNa2O3
  • FormPowder
  • SynonymsCalcinated diatomaceous earth, Diatomite,Diatomaceous Earth, Kieselguhr
  • Weight (g)11340
  • Formula Weight105.988
  • PackagingFiber Drum
  • DOT InformationNot regulated by DOT
  • CAS Number68855-54-9
  • Description545 Filter Aid, Powder, 25 lb
Storage Code Gray

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat