CELLTREAT Scientific Products 229202B Standard-Length Serological Pipette, 2mL, Indv. Wrap, Paper/Plastic, Sterile; 600/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229202B – Item # EW-34567-18
Bold, black ink indicate positive and negative graduations
  • Easily recognize pipette capacity with the color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Sterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes
PRODUCT OPTIONS
$198.00USD / Case of 600
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)2
  • Graduation Interval0.02 mL
  • Increments (mL)0.02
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodeGreen
  • Filter TypeMaterial plug
  • Package Of100 individually wrapped per bag
  • DescriptionStandard-Length Serological Pipet, 2mL, Indv. Wrap, Paper/Plastic, Sterile; 600/Cs
Key Features
  • Easily recognize pipette capacity with the color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Sterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes