CELLTREAT Scientific Products 229212B Standard-Length Serological Pipette, Wide-Tip, 10mL, Indv. Wrap, Paper/Plastic, Sterile; 200/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229212B – Item # EW-34567-35
Bold, black ink indicate positive and negative graduations
  • Easily recognize pipette capacity with the color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Sterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes
PRODUCT OPTIONS
$99.50USD / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)10
  • Graduation Interval0.1 mL
  • Increments (mL)0.1
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodeOrange
  • Filter TypeMaterial plug
  • Package Of50 individually wrapped per bag
  • DescriptionStandard-Length Serological Pipet, Wide-Tip, 10mL, Indv. Wrap, Paper/Plastic, Sterile; 200/Cs
Key Features
  • Easily recognize pipette capacity with the color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Sterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes
  • Features wide tip for use with viscous fluids