CELLTREAT Scientific Products 229224B Open-Ended Serological Pipette, 10mL, Indv. Wrap, Paper/Plastic, Sterile; 200/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229224B – Item # EW-34567-38
Bold, black ink indicate positive and negative graduations
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Perfect for viscous or high particulate fluids—wide openings allow for easier filling and dispensing
PRODUCT OPTIONS
$104.00USD / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)10
  • Graduation Interval0.1 mL
  • Increments (mL)0.1
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodeOrange
  • Filter TypeMaterial plug
  • Package Of50 individually wrapped per bag
  • DescriptionOpen-Ended Serological Pipet, 10mL, Indv. Wrap, Paper/Plastic, Sterile; 200/Cs
Sterile
Key Features
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Perfect for viscous or high particulate fluids—wide openings allow for easier filling and dispensing
More About this Item
These sterile serological pipettes feature bold black positive and negative graduations making them easy to read while flling or dispensing. Color coded stripes and packaging enable quick volume identification.