CELLTREAT Scientific Products 229230A Standard-Length Serological Pipette, 50mL, Indv. Wrap, Plastic/Plastic, Sterile; 90/Cs

Representative image only.
CELLTREAT Scientific Products Mfr # 229230A – Item # EW-34567-42
Bold, black ink indicate positive and negative graduations
  • Easily recognize pipette capacity with the color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Sterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes

PRODUCT OPTIONS

$243.00USD / Case of 90

SPECS & DESCRIPTION

Specifications & Description

  • SterileYes
  • Max Volume (mL)50
  • Graduation Interval0.5 mL
  • Increments (mL)0.5
  • Accuracy (%)2%
  • Body MaterialPolystyrene
  • Color CodePurple
  • Filter TypeMaterial plug
  • Package Of30 individually wrapped per bag
  • DescriptionStandard-Length Serological Pipet, 50mL, Indv. Wrap, Plastic/Plastic, Sterile; 90/Cs

Key Features

  • Easily recognize pipette capacity with the color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Sterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bags
  • Save your controller from vapor damage—each pipette end has a material filter plug
  • Manufactured to ASTM standard E-934 for serological pipettes
  • Choose plastic/plastic wrapped pipettes for reduced particulates, and if wiping the package with alcohol before opening

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat