CELLTREAT Scientific Products Reagent Reservoir, Polystyrene, Sterile, 50 mL, Bulk Packed; 5/Bag, 40 Bags/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229291 – Item # EW-34567-59
Ideal for repetitive liquid transfer with multichannel pipettors
  • Reservoirs are presterilized by gamma irradiation
  • V-shaped angled design promotes funneling of liquid to maximize retrieval and reduce waste
  • Constructed of durable white polystyrene for enhanced solution visibility
  • Bulk packed reservoirs are packed 5 per bag with 40 bags per case for a total of 200 reservoirs per case
PRODUCT OPTIONS
$210.75USD / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)50
  • Wetted MatterialPolystyrene
  • Color CodeWhite
  • Package OfBulk Packed 5/Bag, 40 Bags/Cs
  • DescriptionReagent Reservoir, Polystyrene, Sterile, 50 mL, Bulk Packed; 5/Bag, 40 Bags/Cs
Sterile
Key Features
  • Reservoirs are presterilized by gamma irradiation
  • V-shaped angled design promotes funneling of liquid to maximize retrieval and reduce waste
  • Constructed of durable white polystyrene for enhanced solution visibility
  • Bulk packed reservoirs are packed 5 per bag with 40 bags per case for a total of 200 reservoirs per case
More About this Item
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. Sterile reservoirs are sterilized by gamma irradiation and come individually wrapped or bulk packed.