CELLTREAT Scientific Products Cap for Roller Bottle, Sterile, HDPE, Vented, 0.22 µm; 24/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229391 – Item # EW-01836-36
Cell culture caps—ideal for aseptic biological reactions
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Large knurled cap for easy grip
  • Individually packaged to reduce cross contamination
PRODUCT OPTIONS
$52.40USD / Case of 24
Often Bought With
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Cap IncludedCap Only
  • Cap StyleVented
  • Cap MaterialHDPE
  • PackagingIndividually packaged
  • Special FeaturesFor use with CELLTREAT roller bottles
  • DescriptionCap for Roller Bottle, Sterile, HDPE, Vented, 0.22 µm; 24/Cs
Bio DNase-FreeNonpyrogenicRNase-FreeSterile
Key Features
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
  • Large knurled cap for easy grip
  • Individually packaged to reduce cross contamination
More About this Item
These convenient replacement caps for your tubes, bottles, and flask are sterilized by gamma irradiation and ideal for cell culture applications. Choose from solid or vented caps. Solid caps provide a leakproof seal to keep samples secure. Vented caps feature a 0.22 µm hydrophobic membrane allowing for continuous gas exchange.
Choose the cap the is suitable for use with your CELLTREAT® Scientific Products cell culture product. Roller bottles, flasks, and centrifuge tubes sold separately.