This item is now discontinued. Please browse similar items
CELLTREAT Scientific Products 229448 Micro Centrifuge Tube, 2 mL, Attached Flat Top, Self Standing Bag, Nonsterile; 5000/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229448 – Item # EW-06336-23
LFlat-top cap offers easy one-handed opening and closing
  • Temperature range of -80 to 120°C
  • Feature a frosted marking spot, engraved graduations, and an attached polypropylene flat-top cap
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g15000
  • SterileNo
  • Tube Capacity (mL)2
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Tube Bottom ShapeConical/Skirted
  • Cap IncludedYes
  • Cap StyleFlip-Top
  • Cap MaterialPolypropylene
  • GraduationsYes
  • Packaging1000 per resealable bag; 5 bags per case
  • Special FeaturesNonsterile; skirted bottom
  • DescriptionMicro Centrifuge Tube, 2 mL, Self Standing, Bag, Nonsterile; 5000/Cs
Key Features
  • Temperature range of -80 to 120°C
  • Feature a frosted marking spot, engraved graduations, and an attached polypropylene flat-top cap