CELLTREAT Scientific Products 229478 Bio-Reaction Centrifuge Tube Vent Control Label, Nonsterile; 500/Cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229478 – Item # EW-06336-46
Cell culture caps—ideal for aseptic biological reactions
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCELLTREAT bio-reaction centrifuge tubes
  • DescriptionBio-Reaction Centrifuge Tube Vent Control Label, Nonsterile; 500/Cs
Key Features
  • Sterilized by gamma irradiation
  • DNase-& RNase-free
  • Non-pyrogenic
More About this Item
These convenient replacement caps for your tubes, bottles, and flask are sterilized by gamma irradiation and ideal for cell culture applications. Choose from solid or vented caps. Solid caps provide a leakproof seal to keep samples secure. Vented caps feature a 0.22 µm hydrophobic membrane allowing for continuous gas exchange.
Choose the cap the is suitable for use with your CELLTREAT® Scientific Products cell culture product. Roller bottles, flasks, and centrifuge tubes sold separately.