CELLTREAT Scientific Products 229655 Heavy-Duty Sterile Petri Dishes with Nonvented Lid, 150 x 20 mm; 100/cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229655 – Item # EW-01932-17
Optically clear for improved imaging and observation
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 25 to 28 mL working volume
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHeavy-Duty Sterile Petri Dishes with Nonvented Lid, 150 x 20 mm; 100/cs
Sterile
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 25 to 28 mL working volume
More About this Item
These heavy-duty petri dishes are thicker and stronger for added durability. Stacking rings on the lids provide increased stability.
All dishes are supplied sterile (gamma irradiated) and come packaged 5 dishes/bag.