Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 30 gallon
Representative image only.
Chem-Tainer Mfr # TC1840AP-FA0100-INSTALLED – Item # EW-06929-36
Tanks give you excellent chemical compatibility
PRODUCT OPTIONS
$551.50USD / Each
Often Bought with...
Cole-Parmer ThinTouch™ Nitrile Gloves
$9.00 - $67.50USD / Case of 1000
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene
  • Volume (Gal)30
  • Volume (Liters)114
  • Height (in)40
  • Height (cm)101.6
  • Outside Diameter (OD) (in)18
  • DescriptionConical-Bottom Open-Top Tank with Stand, PE, 30 gallon
Includes
Tank with a pre-drilled drain port and stand.
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.