Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 350 gallon
Representative image only.
Chem-Tainer Mfr # TC4270BP-FA0200-INSTALLED – Item # EW-06929-20
Tanks give you excellent chemical compatibility
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Volume (Gal)350
  • Volume (Liters)1325
  • Height (in)70
  • Height (cm)177.8
  • Outside Diameter (OD) (in)42
  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • DescriptionConical-Bottom Open-Top Tank with Stand, PP, 350 gallon
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.