Chem-Tainer Low-Profile Cylindrical Tank, PE, 90 gallon
Representative image only.
Chem-Tainer Mfr # TC3030AA – Item # EW-06925-65
Low-profile design allows for convenient mixing and filling
  • Resins are FDA compliant
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene
  • Wall thickness (in)0.3125
  • Volume (Gal)90
  • Volume (Liters)341
  • Height (in)30
  • Height (cm)76.2
  • Outside Diameter (OD) (in)30
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • DescriptionLow-Profile Cylindrical Tank with Hinged Cover, PE, 90 gallon
Includes
Cover sold separately.
Key Features
  • Resins are FDA compliant
More About this Item
The self supporting, translucent PE tanks have excellent chemical and impact resistance. Polyethylene covers with a flexible PP hinge and stainless steel (SS) rivets are sold separately.