Chem-Tec 125-TBPNC Adjustable Std Low-Flow Switch for Liquids & Gases, 3.0/500 cc/min, PTFE
Representative image only.
Chem-Tec Mfr # 125-TBPNC – Item # EW-32929-12
$419.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±2%
  • Max Operating Temperature (° F)220
  • Max Operating Temperature (° C)104
  • Communications18" long 22 AWG cable
  • DescriptionAdjustable Std Low-Flow Switch for Liquids & Gases, 3.0/500 cc/min, PTFE
More About this Item
Standard Low-Flow Switches are normally closed (NC) but can be wired for normally open (NO). Typical applications include chemical process and vapor deposition systems; industrial gas lines; pollution control monitoring; atmospheric furnaces; and leak detection.
Choose from a brass switch (32929-10, -14) for pressures up to 1500 psi, or a switch made from PTFE (32929-12, -16) for good chemical compatibility. Wetted materials are brass, epoxy, and Viton for models 32929-10 and -14; PTFE for models 32929-12 and -16. Repeatability is ±2%. Measure 1-¾''W x 2-5/8''H x ¾''D.