Chem-Tec 125-TNO Adjustable Mini Low-Flow Switch for Liquids & Gases, PTFE
Representative image only.
Chem-Tec Mfr # 125-TNO – Item # EW-32929-02
Adjustable set points trigger SPST contacts rated for 70 VA
  • Choose from brass or TFE materials
PRODUCT OPTIONS
$337.00USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±2%
  • Max Operating Temperature (° F)220
  • Max Operating Temperature (° C)104
  • Communications18" long 22 AWG cable
  • DescriptionAdjustable Mini Low-Flow Switch for Liquids & Gases, PTFE
Key Features
  • Choose from brass or TFE materials
More About this Item
Mini Low-Flow Switches are configured to open the SPST contact when flow goes beyond the set point or stops. Typical applications include gas/liquid sampling; chemical injection; pollution control monitoring; atmospheric furnaces; and process systems. Choose from a brass switch (32929-00) for pressures up to 1500 psi or a switch made from PTFE (32929-02) for good chemical compatibility. Wetted materials are brass, epoxy, and Viton® for model 32929-00; PTFE for model 32929-02. Repeatability is ±2%. Measure 1-¼''W x 2-5/8''H x ¾''D.