Chem-Tec Adjustable Industrial Flow Switch for Liquids & Gases, 1/2" NPT(F) Conn
Representative image only.
Chem-Tec Mfr # FAV500BWNC – Item # EW-32929-24
Adjustable set points trigger SPST contacts rated for 70 VA
  • Choose from brass or TFE materials
PRODUCT OPTIONS
$382.00USD / Each
$291.00USD / Each
for an additional
$291.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±2%
  • Max Operating Temperature (° F)220
  • Max Operating Temperature (° C)104
  • Communications18" long 22 AWG cable
  • DescriptionAdjustable Industrial Flow Switch for Liquids & Gases, 1/2" NPT(F) Conn
Key Features
  • Choose from brass or TFE materials
More About this Item
Industrial Flow Switches are normally closed (NC) but can be wired for normally open (NO). Typical applications include lubrication, process, fire control, and cooling systems; heat pumps; hydraulic lifts; and water treatment.
Choose a brass model with a ¼'', 3/8'', ½'', or ¾'' NPT(F) connection. Repeatability is ±2%. Note: Back pressure of 5 psi is required for gas applications. Wetted materials are brass, 316 SS, epoxy, and Viton for all models. Models 32929-20, -22, and -24 measure 3-¼''W x 3-1/4''H x 1''D; model 32929-26 measures 4-¼''W x 4''H x 1''D. Shpg wt 4 lb (1.9 kg).