Chem-Tec FAV-250 Adjustable Industrial Flow Switch for Liquids & Gases, 1/4" NPT(F) Conn
Representative image only.
Chem-Tec Mfr # FAV250BWNC – Item # EW-32929-20
$297.00USD / Each
$272.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±2%
  • Max Operating Temperature (° F)220
  • Max Operating Temperature (° C)104
  • Communications18" long 22 AWG cable
  • DescriptionAdjustable Industrial Flow Switch for Liquids & Gases, 1/4" NPT(F) Conn
More About this Item
Industrial Flow Switches are normally closed (NC) but can be wired for normally open (NO). Typical applications include lubrication, process, fire control, and cooling systems; heat pumps; hydraulic lifts; and water treatment.
Choose a brass model with a ¼'', 3/8'', ½'', or ¾'' NPT(F) connection. Repeatability is ±2%. Note: Back pressure of 5 psi is required for gas applications. Wetted materials are brass, 316 SS, epoxy, and Viton for all models. Models 32929-20, -22, and -24 measure 3-¼''W x 3-1/4''H x 1''D; model 32929-26 measures 4-¼''W x 4''H x 1''D. Shpg wt 4 lb (1.9 kg).