Chemware D1069103 PTFE Boiling Stones/Chips, 1 pound (450 g)
Representative image only.
Chemware Mfr # D1069103 – Item # EW-01199-00
$100.00USD / Pound of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Withstands High Temperature (° F)550
  • DescriptionPTFE Boiling Stones/Chips, 1 pound (450 g)
More About this Item
PTFE boiling stones promote efficient boiling, prevent superheating, and minimize bumping during reflux and speed distillation. Stones will not scratch or abrade glassware. Max temperature is 550°>F (288°>C). Bottle contains 16 oz (450 g).