Chromaflex Jacketed Chromatography Column, 147 mL, 30 cm L x 2.5 cm ID

Representative image only.
Chromaflex Mfr # 420870-3010 – Item # EW-34696-32
PTFE shielded o-rings form a solvent resistant, leak free seal

PRODUCT OPTIONS

$839.00USD / Case of 1

SPECS & DESCRIPTION

Specifications & Description

  • SPE Cartridge Length (mm)300
  • SPE Cartridge Length (cm)30
  • SPE Cartridge Diameter (cm)2.5
  • DescriptionJacketed Chromatography Column, 147 mL, 30 cm L x 2.5 cm ID

Includes

Each column is supplied with two PTFE end fittings with 20 µm porosity HDPE bed supports, five feet (1.5 m) PTFE tubing, two flangeless tubing nuts and two flangeless ferrules. Jacketed columns

More About this Item

Chromaflex® chromatography columns have borosilicate glass construction with PTFE end fittings and HDPE bed supports. Each end fitting has a thin PTFE layer that is curled over the O-ring to form a solvent resistant, leak free seal. These columns can be used with all aqueous buffers. To use with organic solvents order optional PTFE bed supports separately. Columns have a temperature range 0 to 50°C; 1/4-28 flangeless fittings. Use ethylene oxide, N NaOH or 100% ethanol for sterilization (autoclaving is not recommended)

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat