Chromaflex Jacketed Chromatography Column, 491 mL, 100 cm L x 2.5 cm ID

Representative image only.
Chromaflex Mfr # 420870-1010 – Item # EW-34696-36
Unique o-ring design provides a solvent resistant, leak-free seal
  • Constructed of borosilicate glass 3.3, PTFE, and HDPE for exceptional chemical resistance
  • Jacketed column features a glass barrel surrounded by a water jacket
  • Column has a 491 mL capacity

PRODUCT OPTIONS

$1,098.00USD / Case of 1
Include Accessories Today....

SPECS & DESCRIPTION

Specifications & Description

  • Column Length (cm)100
  • Column Length (mm)1000
  • Column Inside Diameter (ID) (mm)2.5
  • Max Pressure (PSI)75
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° C)50
  • Total Volume Capacity (mL)491
  • MethodologyJacketed Chromatography Column
  • DescriptionJacketed Chromatography Column, 491 mL, 100 cm L x 2.5 cm ID

Includes

two PTFE end fittings with 20 µm porosity HDPE bed supports, screw caps, five feet (1.5 m) PTFE tubing, two flangeless tubing nuts and two flangeless ferrules, water jacket, and two jacket sealing rings.

Key Features

  • Constructed of borosilicate glass 3.3, PTFE, and HDPE for exceptional chemical resistance
  • Jacketed column features a glass barrel surrounded by a water jacket
  • Column has a 491 mL capacity

More About this Item

CHROMAFLEX® chromatography columns feature uniquely designed PTFE shielded o-rings to form a solvent resistant, leak-free seal. Each end fitting has a thin PTFE layer that is curled over the o-ring. Columns can be used with all aqueous buffers without modification and with organic solvents when the optional PTFE bed supports are installed in the end fittings and flow adapters. Columns also feature borosilicate glass barrel construction with PTFE end fittings and HDPE bed supports. Recommended sterilization is with ethylene oxide, 2N NaOH or 100% ethanol (autoclaving is not recommended).

Choose from standard or jacketed columns. Jacketed columns feature a water jacket around the barrel to provide thermal control of temperature-sensitive chromatographic separations. The jacket also acts as a safety shield.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat