Chromatography Research Supplies Puresep-P 230650 Puresep™ P Septa, 7/16" (11.0 mm); 50/Pk
Representative image only.
Chromatography Research Supplies Mfr # 230650 – Item # EW-01994-02
Designed for enhanced bond and durability
  • Constructed of silicone rubber with a stable layer of polyimide film
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.4375
  • DescriptionSepta, 7/16\" (11.0 mm); 50/pack
Key Features
  • Constructed of silicone rubber with a stable layer of polyimide film
More About this Item
PURESEP-P™ septa are made of silicone rubber with a stable layer of polyimide film; withstand temperatures to 572°F (300°C).