CINC SPARE PART KIT V2 Spare Parts Kit, contains common wear parts
Representative image only.
CINC Mfr # SPARE PART KIT V2 – Item # EW-17315-08
Ideal for maintenance of your Centrifugal Multipurpose Separator/Extractors
  • Optimize performance of your CINC Centrifugal Separators
$630.00USD / Set of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSpare Parts Kit, contains common wear parts
Key Features
  • Optimize performance of your CINC Centrifugal Separators
More About this Item
The spare parts kit, model 17315-08, includes:
3-6-32 Socket Head Caps Screw 3/8 Long
3-8-32 Socket Head Caps Screw 3/8 Long
1-Bottom Plate O-ring, Parker 2-038, PTFE
1-Housing O-ring, Parker 2-041, PTFE
2-Rotor Head and Sleeve O-ring, Parker 2-033, PTFE
Encapsulated Viton
1-Seal Ring
1-Motor - Rotor Flex Sleeve Coupling
1-Bearing Retainer Ring
2-Rotor Bearing
1-Bearing Lock Nut
1-Bearing Preload Spring
1-Upper Seal
1-Low Mix Seal if Applicable
1-Spacer, Inner
1-Spacer, Outer