Clift Industries 2901-005 Hf Acid Eater Spill Kit; 5 Gal.; 1 Pack/case
Representative image only.
Clift Industries Mfr # 2901-005 – Item # EW-86295-04
Neutralizes Hydrofluoric Acid into safe Organic salts
  • 2 year warranty
$302.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHf Acid Eater Spill Kit; 5 Gal.; 1 Pack/case
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Unlike caustic soda and soda ash, which forms poisonous bi-fluoride, HF Acid Eater completely destroys the hydrofluoric acid polymer. This neutralizer has a 1:1 neutralization ratio as apposed to the 12:1 ratio of caustic soda. pH indicator additive eliminates the need for additional pH strips taking out the guess work. HF Acid Eater is nonhazardous and noncorrosive, and is excellent for a variety of uses and applications. Apply on horizontal, vertical, and perforated surfaces. Also neutralizes vapors and reduces odors.