This item is now discontinued. Please browse similar items
Cole-Parmer 0.01 to 100 mTorr Pressure/Vacuum Controller for Pirani-Type Sensor
Representative image only.
Cole-Parmer Item # EW-68801-20
Customize to your needs
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% of reading or ±0.03 mTorr
  • Unit Of MeasurementmTorr
  • Display4.5-digit LED
  • Battery Type115 VAC, 50/60 Hz
  • Electrical Connections5.5-ft (1.7-m) power cord
  • Switch output3 A at 115 VAC (noninductive)
  • Output1 mV/digit, SPDT
  • Resolution0.01 mTorr
  • Description0.01 to 100 mtorr Pressure/Vacuum Controller for Pirani-Type Sensor
Key Features
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
More About this Item
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate.
These sensors are suited for use with mercury and oil pumps.
Pirani type sensors offer enhanced stability and an automatic temperature compensation network. The 304 stainless steel sensors operate at half the
temperature of standard thermopile sensors. Use for high-vacuum applications.
Diaphragm type sensor offers quick response—less than 400 milliseconds full-scale. Select a diaphragm type sensor made from 316 stainless steel or silicon based on your application.