Cole-Parmer 0/50 Percent Isopropyl Alcohol Hydrometer
Representative image only.
Cole-Parmer Item # EW-08299-71
  • NIST Traceable
  • Metal Ballast
  • Individually Serialized
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")10 1/5
  • Hydrometer FormPlain
  • Alcohol Range (Min) (%)0
  • Alcohol Range (Max) (%)50
  • Division0.01
  • Accuracy±1%
  • Length (mm)260
  • Description 0/50 Percent Isopropyl Alcohol Hydrometer
  • Warranty5 years
5 Year Warranty
Key Features
  • NIST Traceable
  • Metal Ballast
  • Individually Serialized
  • 5 year warranty
More About this Item
For determining proof of alcohol percent, all hydrometers are made according to the Bureau of Internal Revenue, US Treasure Department. The tolerances listed for these hydrometers are those outlined in Circular 555 of the National Bureau of Standards, Testing of Hydrometers. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Equivalent to H-B Instrument 618094000.