Cole-Parmer 1.100/1.150 Specific Gravity ASTM 127H Hydrometer for Heavy Liquids
Representative image only.
Cole-Parmer Item # EW-08297-84
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Conform to ASTM Specifications
  • Metal Ballast
  • Individually Serialized
  • 5 year warranty
PRODUCT OPTIONS
$52.36USD / Each
$61.60USD / Each
$176.00 - $249.00USD / Each
for an additional
$176.00USD / Each
for an additional
$249.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Hydrometer FormPlain
  • Length (\")10 1/5
  • Liquids Heavier Than WaterYes
  • Specific Gravity Range (Min) (%)1.1
  • Specific Gravity Range (Max) (%)1.15
  • Division0.001
  • Accuracy±0.001
  • ASTM number127H
  • DescriptionPlain-form Glass Hydrometer, ASTM 127H, 1.100-1.150 Sg/0.0005
  • Warranty5 years
5 Year Warranty
Key Features
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Conform to ASTM Specifications
  • Metal Ballast
  • Individually Serialized
  • 5 year warranty
More About this Item
These hydrometers conform to the accuracy requirements for ASTM hydrometers. Scales are affixed with clear epoxy. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Equivalent to H-B Instrument 618913600.