Cole-Parmer 150134 High-Purity #12 PP Housing for 10" 222 SOE O-Ring Cartridges
Representative image only.
Cole-Parmer Item # EW-01508-24
Filter housing is ideal for in-line process filtration systems
  • Constructed of durable polypropylene (PP)
$103.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Length (in)10
  • DescriptionHigh-Purity #12 PP Housing for 10" 222 SOE O-Ring Cartridges
Key Features
  • Constructed of durable polypropylene (PP)
More About this Item
This #12 housing fits submicronic cartridges with 222 O-ring sealing caps. The pure PP housing contains no filters, lubricants, or plasticizers. Housing features an O-ring sealed PP drain plug in the sump with a second vent for differential pressure.
Maximum temperature is 100°>F (38°>C); maximum pressure is 100 psi. Inlet/outlet ports are 3/4" NPT(F); length is 14-3/8". Includes VITON O-ring. Shpg wt 3 lb (1.4 kg).