Cole-Parmer 155155-43 Double open end granular activated carbon cartridge; 20 µm, 10"L
Representative image only.
Cole-Parmer Item # EW-01508-92
$19.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaActivated Carbon
  • Micron Rating20
  • Flow Rate1 GPM
  • Filter Length (in)10
  • Max Temperature (° F)125
  • Max Temperature (° C)51.7
  • DescriptionDouble open end granular activated carbon cartridge; 20 µm, 10"L
More About this Item
Granular Activated Carbon Cartridges remove odors and chlorine taste from water. Approximately 10-oz of granular carbon is packed in the plastic tube. End caps have filters to protect carbon. Use at 1 GPM or lower. For cold water only. May initially contain leachable carbon dust; flush system well after installation. Maximum temperature is 125°>F (52°>C); maximum differential pressure is 70 psid**. Shpg wt 2 lb (1.0 kg).
** For optional performance, change cartridges at 25 psid