Cole-Parmer 155395-03 Polypropylene felt filter bag, 10"L, 200 µm
Representative image only.
Cole-Parmer Item # EW-01516-12
$7.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating200
  • Flow Rate20 GPM
  • Filter Length (in)10
  • DescriptionPolypropylene felt filter bag, 10"L, 200 µm
More About this Item
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.