Cole-Parmer 155751-43 Double open end polypropylene sediment cartridge; 25 µm, 10 inch length
Representative image only.
Cole-Parmer Item # EW-01512-28
Polypropylene fiber medium is thermally bonded to a rigid core for added durability
PRODUCT OPTIONS
$16.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating25
  • Flow Rate2 GPM at <2 psid
  • Filter Length (in)10
  • DescriptionDouble open end polypropylene sediment cartridge; 25 µm, 10 inch length
More About this Item
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 175°F (79°C).
Note: For optimal performance, change cartridges at 25 psid.