Cole-Parmer 200 µL Universal Grad Tip, Sterile (Individually Wrapped); 400 Tips/Box, 5 Box/Case
Representative image only.
Cole-Parmer Item # EW-15977-66
Broad range of standard and filter pipette tips
  • FDA approved special medical grade virgin polypropylene with high transparency
  • Sterile tips are free of DNase, RNase, Human DNA, Pyrogen
  • Tips load and eject easily for reduced hand fatigue
  • Compatible with wide range of pipettes
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (µL)200
  • Autoclavableyes
  • Package Of2000/Cs
  • Description200 uL Universal Grad tip, Sterile (Individually Wrapped), 5x400 Tips/Case
Sterile
Key Features
  • FDA approved special medical grade virgin polypropylene with high transparency
  • Sterile tips are free of DNase, RNase, Human DNA, Pyrogen
  • Tips load and eject easily for reduced hand fatigue
  • Compatible with wide range of pipettes