Cole-Parmer 225-XL Disc Clips for Roto-Torque Rotator, 25-40 mm
Representative image only.
Cole-Parmer Item # EW-07637-70
$23.20USD / Dozen of 1
$58.00USD / Dozen of 1
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDisc Clips for Roto-Torque Rotator, 25-40 mm
More About this Item
Parts and accessories to meet your shaking mixer application needs.
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
1 Question | 1 Answer
Displaying question 1
  • Q:
    Hello,
    Will the 25-40 mm Disc Clips work for holding 50 mL conical tubes?
    Thank you.
    Asked on 2/7/2012 by Natasha from San Diego, CA
    1 answer
    • STAFF EXPERT
      A:
      Yes, most 50ml conical tubes are between 26-32mm in diameter. So these would be the appropriate clips.
      Answered on 2/8/2012 by Mike from Tech Support
Displaying question 1
Do you have a question about this product? 