Cole-Parmer 227670 Replacement Robotic Stoppers for BOD Bottles
Representative image only.
Cole-Parmer Item # EW-03660-03
Enhance the versitility of your Wheaton BOD Bottles
  • Choose from a wide range of BOD bottle accessories
PRODUCT OPTIONS
$101.00USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Compatible WithBOD bottles
  • DescriptionReplacement Robotic Stoppers for BOD Bottles
Key Features
  • Choose from a wide range of BOD bottle accessories
More About this Item
Model 03660-09 cap and insert is for use with 03660-15 BOD bottles. Model 03660-12 rubber bottle leash holds the stopper to the bottle to avoid misplacing the stopper or breakage.