Cole-Parmer 255001-03 Polypropylene absolute-rated high-efficiency filter bags; 10"L, 1 µm
Representative image only.
Cole-Parmer Item # EW-01516-40
Conveniently collect all filtered particles within the bag
  • Choose from a wide range of bag materials suitable for your application
PRODUCT OPTIONS
$14.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating1
  • Flow Rate15 GPM
  • Filter Length (in)10
  • DescriptionPolypropylene absolute-rated high-efficiency filter bags; 10"L, 1 µm
Key Features
  • Choose from a wide range of bag materials suitable for your application
More About this Item
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.