Cole-Parmer 255016-03 Nylon monofilament filter bag; 10"L, 800 µm
Representative image only.
Cole-Parmer Item # EW-01516-76
$16.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaNylon
  • Micron Rating800
  • Flow Rate35 GPM
  • Filter Length (in)10
  • DescriptionNylon monofilament filter bag; 10"L, 800 µm
More About this Item
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.