Cole-Parmer 255026-03 Polypropylene absolute-rated high-efficiency filter bags; 18-1/2"L, 5 µm
Representative image only.
Cole-Parmer Item # EW-01516-84
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating5
  • Flow Rate35 GPM
  • Filter Length (in)18.5
  • DescriptionPolypropylene absolute-rated high-efficiency filter bags; 18-1/2"L, 5 µm
More About this Item
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.