Cole-Parmer 255187-03 Size 3 polypropylene felt filter bag; 10 µm
Representative image only.
Cole-Parmer Item # EW-01519-72
Attach bags onto any open system drain or sump-return to collect solids
  • Solids are contained within the bag for easy disposal
$4.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating10
  • Flow Rate10 GPM
  • Filter Length (in)8
  • Filter Diameter (in)4.125
  • Filtration Area (ft²)0.5
  • Filtration Area (in²)72
  • Filtration Area (m²)0.046
  • DescriptionSize 3 polypropylene felt filter bag; 10 µm
Key Features
  • Solids are contained within the bag for easy disposal
More About this Item
An improvement over tie-on designs—the head and support basket make the full filter bag surface area available for use. Filter bags include a galvanized steel ring sewn into the top that
securely holds the bag to the head. Polypropylene bags are glazed to reduce fiber migration and have welded seams for high strength. All bag designs are suitable to 100°F (38°C).