Cole-Parmer 25H-1705-CP Vent Filter, 40 mm, 0.2 um, Autoclave; 1 Ea
Representative image only.
Cole-Parmer Item # EW-06063-14
Ideal for hydrophobic venting and solvent filtration applications
  • Complies with USP Class VI standards
  • Autoclavable up to 10 times
PRODUCT OPTIONS
$14.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPTFE
  • Housing MaterialPolypropylene (PP)
  • Pore Size (µm)0.2
  • Filter Diameter (mm)40
  • DescriptionCarboy Vent Filter, 40 mm, 0.2 um, Autoclavable; 1 Ea
Key Features
  • Complies with USP Class VI standards
  • Autoclavable up to 10 times
More About this Item
The non-sterile vent filters feature a polypropylene housing with a 0.2 µm hydrophobic PTFE membrane—ideal for maintaining purified water storage in carboys or for sterile venting during a slow exhaust/liquid autoclave cycle of EZgrip™ carboys. They can withstand up to 10 autoclave cycles, feature a stepped hose barb, and are available in 40 mm gamma safe or 50 mm autoclave only membrane sizes.