YSI 5776 High-Sensitivity DO Membrane Kit
Representative image only.
YSI Mfr # 098095 – Item # EW-05522-10
Available in standard or high-sensitivity versions
  • Each kit contains 30 membranes and 5.25 g of KCL crystals.
  • 1 year warranty
PRODUCT OPTIONS
$76.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Membranes
  • DescriptionHigh-Sensitivity DO Membrane Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • Each kit contains 30 membranes and 5.25 g of KCL crystals.
  • 1 year warranty
More About this Item
Use the standard kit (05522-00) for general laboratory DO needs; or choose the high-sensitivity kit (05522-10) for oxygen levels <= 5 mg/L at 15°C or below.