Digi-Sense Traceable® Digital Thermohygrometer with Dew Point, Memory Card and Calibration
Representative image only.
Digi-Sense Item # EW-37804-21
Easily transfer data using an SD card from your meter to a PC
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • SD memory card stores up to 5.9 million readings
  • Adjustable data logging sampling time from 1 second to 12 hours
  • Maximum and minimum readings can be recalled or stored for later use
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)4.25
  • Width (in)3.5
  • Depth (InchShort)1.25
  • DescriptionTraceable® Digital Thermohygrometer with Dew Point, Memory Card and Calibration
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Get accurate, reliable measurements with included NIST-traceable calibration
  • SD memory card stores up to 5.9 million readings
  • Adjustable data logging sampling time from 1 second to 12 hours
  • Maximum and minimum readings can be recalled or stored for later use
  • 1 year warranty
More About this Item
This thermohygrometer records data onto a standard SD memory card instead of built-in memory — the advantage being that when the memory on the SD card is full, the SD card can be simply exchanged out for unlimited data storage. The data is provided in a text report that includes date, time of day, and readings of sensors, and may be sent to any spreadsheet or database.
The unique 4-line display simultaneously shows ambient temperature, humidity, dew point, and time of day. An audible and visual alarm alerts you when temperature, humidity, or dew point levels are higher or lower than the programmable set points. User-settable parameters include date, time, sampling time, temperature unit of measure, high/low alarm set points, and auto power off.
Traceable® products are calibrated to NIST-traceable standards and ready to use as soon as you receive it. Each product goes through an intense vetting process before it can carry the Traceable name, and it includes NIST-traceable certification. This means that a 17025-accredited calibration test lab has confirmed that the product meets or exceeds all stated specifications, saving you both time and money by not having to calibrate separately.
NIST-traceable calibration also helps you meet quality standards and regulatory compliance for ISO, FDA, cGMP, VFC, CAP, CLIA and Joint Commission requirements.