Cole-Parmer 6761P Replacement Impactor for Small Grinding Set 04576-34, PC Encapsulated Steel; 1/EA
Representative image only.
Cole-Parmer Item # EW-04577-74
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Polycarbonate encapsulated steel impactor
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
PRODUCT OPTIONS
$119.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPC encapsulated steel
  • Compatible WithSmall grinding vial set 04576-34
  • DescriptionReplacement Impactor for Small Grinding Set 04576-34, PC Encapsulated Steel; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Polycarbonate encapsulated steel impactor
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.