Cole-Parmer 6771 Grinding Vial Set, Small Vial, 0.5 to 4 mL, PC Cylinder, Low-Chrome Steel End Plugs and Impactor

Representative image only.
Cole-Parmer Item # EW-04576-35
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Specifically designed to grind electronic components for RoHS/WEEE compliance testing
  • Withstands liquid nitrogen temperatures

PRODUCT OPTIONS

$452.00USD / Pkg of 1

SPECS & DESCRIPTION

Specifications & Description

  • Use with GeneratorsFreezer/Mill® cryogenic grinders 04500-39 to -42 and 04577-77, -92 to -94
  • Min Sample Size (mL)0.5
  • Min Sample Size (g)0.5
  • Max Sample Size (mL)4
  • Max Sample Size (g)5
  • MaterialPC cylinder, low-chrome steel end plugs and impactor
  • DescriptionGrinding Vial Set, Small Vial, 0.5 to 4 mL, PC Cylinder, Low-Chrome Steel End Plugs and Impactor

Includes

One set of stainless steel end plugs, one low-chrome steel impactor, and four polycarbonate center cylinder.

Key Features

  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Specifically designed to grind electronic components for RoHS/WEEE compliance testing
  • Withstands liquid nitrogen temperatures
  • Transparent center to view progress
  • Sample capacity of 0.1 to 5 g

More About this Item

Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.

The closed grinding vial prevents sample cross-contamination, retains volatiles, and protects hazardous and critical samples. End plugs and impactor are made of Cr-free ASTM 06 steel to eliminate chromium contamination during grinding. Low-chrome steel end plugs and impactor are autoclavable and reusable; polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent. Grinding vial set is compatible with CG-200 Freezer/Mill® cryogenic grinder. For use with CG-400, CG-450, and CG-500 Freezer/Mill® cryogenic grinders, order multivial holder 04500-51. Easily remove end plugs using extractor 04550-46.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat