Cole-Parmer 6771P Replacement Impactor for Small Grinding Vial Set 04576-35, Low-Chrome Steel; 1/EA
Representative image only.
Cole-Parmer Item # EW-04577-76
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Made of Cr-free ASTM 06 steel to eliminate chromium contaminationduring grinding
  • Specifically designed for grinding electronic components for RoHS/WEEE compliance testing
  • Autoclavable and reusable
PRODUCT OPTIONS
$104.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLow-chrome steel
  • Compatible WithSmall grinding vial set 04576-35
  • DescriptionReplacement Impactor for Small Grinding Vial Set 04576-35, Low-Chrome Steel; 1/EA
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Made of Cr-free ASTM 06 steel to eliminate chromium contaminationduring grinding
  • Specifically designed for grinding electronic components for RoHS/WEEE compliance testing
  • Autoclavable and reusable
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.