Cole-Parmer 6781E Replacement End Plugs for Small Grinding Vial Set 04577-80, SS; 2/PK
Representative image only.
Cole-Parmer Item # EW-04577-78
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Ideal for when contact with plastic is not recommended
  • Autoclavable and reusable
PRODUCT OPTIONS
$260.00USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless steel
  • Compatible WithSmall grinding vial set 04577-80
  • DescriptionReplacement End Plugs for Small Grinding Vial Set 04577-80, SS; 2/PK
Includes
10 O-rings.
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Ideal for when contact with plastic is not recommended
  • Autoclavable and reusable
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.