Cole-Parmer 6803 Grinding Vial Set, Large Vial, 1 to 50 mL, SS Cylinder, End plugs with O-Rings, and Impactor

Representative image only.
Cole-Parmer Item # EW-61043-10
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Ideal for when contact with plastic is not recommended
  • Easy to clean and reusable

PRODUCT OPTIONS

$913.00USD / Pkg of 1

SPECS & DESCRIPTION

Specifications & Description

  • Use with GeneratorsFreezer/Mill® cryogenic grinders 04500-40 to -42 and 04577-92 to -99
  • Min Sample Size (mL)1
  • Min Sample Size (g)5
  • Max Sample Size (mL)50
  • Max Sample Size (g)50
  • MaterialSS cylinder, end plugs with O-ring, and impactor
  • DescriptionGrinding Vial Set, Large Vial, 1 to 50 mL, SS Cylinder, End plugs with O-Rings, and Impactor

Includes

Ones et of stainless steel (SS) end plugs, one SS impactor, one SS cylinder, and 10 O-rings.

Key Features

  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Ideal for when contact with plastic is not recommended
  • Easy to clean and reusable
  • Sample capacity of 5 to 50 g

More About this Item

Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.

The closed grinding vial prevents sample cross-contamination, retains volatiles, and protects hazardous and critical samples. All stainless steel material makes it ideal for grinding hard samples like bone and circuit boards. End plugs, impactor, and cylinder are autoclavable and reusable. Grinding vial set is compatible with CG-200, CG-400, CG-450, and CG-500 Freezer/Mill® cryogenic grinders. Easily remove end plugs using extractor 04550-50.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat