Cole-Parmer 6883E Replacement End Plugs for Mid-Size Grinding Vial Set 61043-12, Low-Chrome Steel; 2/PK
Representative image only.
Cole-Parmer Item # EW-61034-64
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Made of Cr-free ASTM 06 steel to eliminate chromium contaminationduring grinding
  • Specifically designed for grinding electronic components for RoHS/WEEE compliance testing
  • Autoclavable and reusable
PRODUCT OPTIONS
$380.00USD / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLow-chrome steel
  • Compatible WithMid-size grinding vial set 61043-12
  • DescriptionReplacement End Plugs for Mid-Size Grinding Vial Set 61043-12, Low-Chrome Steel; 2/PK
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Made of Cr-free ASTM 06 steel to eliminate chromium contaminationduring grinding
  • Specifically designed for grinding electronic components for RoHS/WEEE compliance testing
  • Autoclavable and reusable
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.