This item is now discontinued. View the direct replacement: EW-24501-08 or browse similar items
Cole-Parmer A100 Adjustable-Volume Pipettor; 100 to 1000 uL
Representative image only.
Cole-Parmer Item # EW-15970-65
Fully autoclavable with enhanced UV resistance
  • Ergonomic, comfortable, and light weight
  • Superior accuracy and precision
  • Locking counter prevents accidental volume fluctuations
  • Easily identify your pipette using detachable color-coded drops
  • Reverse pipetting option ensures accurate, precise aspirating and dispensing of viscous liquids
SPECS & DESCRIPTION
Specifications & Description
  • AutoclavableYes
  • Max Volume (µL)1000
  • Min Volume (µL)100
  • Increments (µL)5
  • Accuracy (%)±0.6
  • Precision (%)±0.20
  • Number Of Channels1
  • Tip EjectorYes
  • Color CodeYes
  • DescriptionAdjustable-Volume Single Channel Pipettor
Includes
Pipettor, calibration/disassembling tool, color coding drops and remover tool, silicon grease, and sample tips
Key Features
  • Ergonomic, comfortable, and light weight
  • Superior accuracy and precision
  • Locking counter prevents accidental volume fluctuations
  • Easily identify your pipette using detachable color-coded drops
  • Reverse pipetting option ensures accurate, precise aspirating and dispensing of viscous liquids
  • Calibration stability guarantees accuracy of dispense volume even after multiple autoclaving and pipetting
  • Enhanced UV resistance provides added protection from biohazardous elements
  • Universal tip compatibility to use with wide range of tips up to 1000μL
  • Meets DIN EN ISO 8655 standard
More About this Item
Cole-Parmer A100-series Single Channel Adjustable-Volume Pipettors have been ergonomically designed to the most rigorous standards for accuracy, precision and comfort. Pipettor’s user-friendly handle design with soft grip gives ease of operation and reduces fatigue. Its’ light weight and low plunger force minimizes RSI (Repetitive Strain Injury). Sturdy PVDF tip cone ensures a high resistance against chemicals. Blow out system ensures last drop dispensing thus preventing critical sample loss. Innovative soft grip protects heat transfer to internal components ensuring accurate operation.