Cole-Parmer Accumet-Compatible Replacement pH/ATC Probe
Representative image only.
Cole-Parmer Item # EW-59500-81
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsAll-in-one pH/ATC
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference ElectrolyteAg/AgCl
  • Temperature CompensationYes
  • ATC ConnectorAccumet compatible
  • ConnectorBNC
  • DescriptionAccumet-Compatible Replacement pH/ATC Probe
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This is an epoxy body, single junction, Ag/AgCl reference, 10K thermistor ATC (compatible with accumet® models), refillable pH electrode. Dimensions are 12mm x 110mm.