Cole-Parmer Acetone, ACS Grade, Amber Glass Bottle, 2.5 L

Representative image only.
Cole-Parmer Item # EW-88406-13
Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
  • Aldehyde (as HCHO): max 0.002%
  • Assay: min 99.5%
  • Color (APHA): max 10
  • Isopropyl Alcohol: max 0.05%
  • Methanol (Methyl Alcohol): max 0.05%
  • Residue after evaporation (Non-Volatile Matter): max 0.001%
  • Solubility in Water: To Pass Test
  • Substances Reducing Permanganate: To Pass Test
  • Titrable Acid: max 0.0003 meq/g
  • Titrable Base: max 0.0006 meq/g
  • Water (H2O): max 0.5%

PRODUCT OPTIONS

$137.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical Formula(CH3)2CO
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms2-Propanone, Pyroacetic Ether, 2-Propanone, Dimethyl Ketone
  • Volume (mL)2500
  • Formula Weight58.07
  • PackagingAmber Glass Bottle
  • DOT InformationAcetone; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolFlammable
  • CAS Number67-64-1
  • UN NumberUN1090
  • Shelf Life (months)60
  • DescriptionAcetone, ACS Grade, Amber Glass Bottle, 2.5 L
FlammableStorage Code Red

Key Features

  • Quality and consistency your lab can count on
  • Aldehyde (as HCHO): max 0.002%
  • Assay: min 99.5%
  • Color (APHA): max 10
  • Isopropyl Alcohol: max 0.05%
  • Methanol (Methyl Alcohol): max 0.05%
  • Residue after evaporation (Non-Volatile Matter): max 0.001%
  • Solubility in Water: To Pass Test
  • Substances Reducing Permanganate: To Pass Test
  • Titrable Acid: max 0.0003 meq/g
  • Titrable Base: max 0.0006 meq/g
  • Water (H2O): max 0.5%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Please note this product may require additional documentation/verification to purchase. This may include business identification and an authorized purchaser form:
Acceptable business identification
State issued document with your business information

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat